Assay Design



Gene Transcripts

Gene Symbol
Ccdc60
Entrez Gene ID
269693
Gene Name
coiled-coil domain containing 60
Gene Aliases
AU018954, C130098D09
Chromosome Location
Chr.5: 116125581 - 116288998 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177759.3 NP_808427.1 3 - 4 789 80
GenBank mRNA AK082047.1 - 3 - 4 790 80
BC075662.1 - 3 - 4 768 80
BC150892.1 - 3 - 4 579 80

View Full Product Details