Assay Design
Gene Transcripts
- Gene Symbol
- Alkbh2
- Entrez Gene ID
-
231642
- Gene Name
- alkB, alkylation repair homolog 2 (E. coli)
- Gene Aliases
-
9530023G02, AU016977, Abh2, mABH2
- Chromosome Location
-
Chr.5: 114123926 - 114130546 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_175016.2 |
NP_778181.2 |
1 - 2 |
305 |
83 |
GenBank mRNA |
BC062188.1 |
- |
1 - 2 |
305 |
83 |
View Full Product Details