Assay Design



Gene Transcripts

Gene Symbol
Oas3
Entrez Gene ID
246727
Gene Name
2'-5' oligoadenylate synthetase 3
Gene Aliases
Oasl10
Chromosome Location
Chr.5: 120753098 - 120777659 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145226.2 NP_660261.1 14 - 15 3299 92
GenBank mRNA AB067534.1 - 14 - 15 3299 92
AF453830.1 - 14 - 15 3291 92
BC016461.1 - 7 - 8 1397 92
BC141055.1 - 14 - 15 3292 92

View Full Product Details