Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016698.2 | NP_057907.2 | 7 - 8 | 1604 | 64 |
GenBank mRNA | AB026621.1 | - | 7 - 8 | 1603 | 64 |
AK004579.1 | - | 7 - 8 | 1599 | 64 | |
AK034953.1 | - | 7 - 8 | 1603 | 64 | |
AK129106.1 | - | 7 - 8 | 1592 | 64 | |
AK146728.1 | - | 7 - 8 | 1604 | 64 | |
AK149972.1 | - | 7 - 8 | 1605 | 64 | |
AK156502.1 | - | 7 - 8 | 1601 | 64 | |
AK165617.1 | - | 7 - 8 | 1604 | 64 | |
AK168523.1 | - | 7 - 8 | 1599 | 64 | |
AK172065.1 | - | 7 - 8 | 1602 | 64 | |
AK218316.1 | - | 1 - 2 | 96 | 64 | |
BC010342.1 | - | 7 - 8 | 1313 | 64 |