Assay Design



Gene Transcripts

Gene Symbol
Klk5
Entrez Gene ID
68668
Gene Name
kallikrein related-peptidase 5
Gene Aliases
1110030O19Rik
Chromosome Location
Chr.7: 43834157 - 43851181 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026806.2 NP_081082.1 2 - 3 387 64
GenBank mRNA AK003996.1 - 2 - 3 388 64
BC139475.1 - 2 - 3 388 64
BC139478.1 - 2 - 3 388 64

View Full Product Details