Assay Design
Gene Transcripts
- Gene Symbol
- Bri3
- Entrez Gene ID
-
55950
- Gene Name
- brain protein I3
- Gene Aliases
-
I3
- Chromosome Location
-
Chr.5: 144244437 - 144446757 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK049027.1 |
- |
3 - 4 |
527 |
68 |
View Full Product Details