Assay Design
Gene Transcripts
- Gene Symbol
- Serpine1
- Entrez Gene ID
-
18787
- Gene Name
- serine (or cysteine) peptidase inhibitor, clade E, member 1
- Gene Aliases
-
PAI-1, PAI1, Planh1
- Chromosome Location
-
Chr.5: 137061504 - 137072272 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_008871.2 |
NP_032897.2 |
8 - 9 |
1314 |
70 |
GenBank mRNA |
BC054091.1 |
- |
8 - 9 |
1311 |
70 |
View Full Product Details