Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198109.4 | NP_932777.2 | 3 - 4 | 240 | 78 |
GenBank mRNA | AB099287.1 | - | 3 - 4 | 275 | 78 |
AK089308.1 | - | 3 - 4 | 244 | 78 | |
AK155557.1 | - | 3 - 4 | 265 | 78 | |
AK165380.1 | - | 2 - 3 | 203 | 78 | |
AK171210.1 | - | 3 - 4 | 289 | 78 | |
AK171556.1 | - | 3 - 4 | 255 | 78 | |
AK173144.1 | - | 3 - 4 | 262 | 78 |