Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_198109.4 | NP_932777.2 | 6 - 7 | 495 | 54 |
GenBank mRNA | AB099287.1 | - | 6 - 7 | 530 | 54 |
AK089308.1 | - | 6 - 7 | 499 | 54 | |
AK155557.1 | - | 6 - 7 | 520 | 54 | |
AK165380.1 | - | 5 - 6 | 460 | 54 | |
AK171210.1 | - | 6 - 7 | 544 | 54 | |
AK171556.1 | - | 6 - 7 | 510 | 54 |