Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033229.3 | NP_001028401.2 | 2 - 3 | 430 | 95 |
GenBank mRNA | AK142439.1 | - | 2 - 3 | 407 | 95 |
AK163083.1 | - | 2 - 3 | 406 | 95 | |
AK168839.1 | - | 2 - 3 | 407 | 95 | |
BC139248.1 | - | 2 - 3 | 430 | 95 | |
BC139250.1 | - | 2 - 3 | 430 | 95 |