Assay Design



Gene Transcripts

Gene Symbol
Kif20b
Entrez Gene ID
240641
Gene Name
kinesin family member 20B
Gene Aliases
33cex, B130024C23, C330014J10Rik, Mphosph1, magoo
Chromosome Location
Chr.19: 34922338 - 34975750 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_183046.1 NP_898867.1 23 - 24 4156 116
GenBank mRNA AK145779.1 - 11 - 12 2399 116
AK162252.1 - 4 - 5 117
AY259532.1 - 15 - 16 2933 116
BC036147.1 - 5 - 6 1578 116
BC048954.1 - 4 - 5 388 116
BC151061.1 - 23 - 24 4002 116
BC172148.1 - 23 - 24 4002 116

View Full Product Details