Assay Design



Gene Transcripts

Gene Symbol
Cyp2c55
Entrez Gene ID
72082
Gene Name
cytochrome P450, family 2, subfamily c, polypeptide 55
Gene Aliases
2010318C06Rik
Chromosome Location
Chr.19: 39006938 - 39042687 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028089.3 NP_082365.1 6 - 7 988 71
GenBank mRNA AK008580.1 - 6 - 7 988 71
AY206875.1 - 6 - 7 961 71
BC010824.1 - 6 - 7 976 71

View Full Product Details