Assay Design



Gene Transcripts

Gene Symbol
Osbp
Entrez Gene ID
76303
Gene Name
oxysterol binding protein
Gene Aliases
1110018F06Rik, AW559088, mKIAA4220
Chromosome Location
Chr.19: 11965844 - 11994114 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033174.1 NP_001028346.1 3 - 4 915 83
GenBank mRNA BC107327.1 - 3 - 4 649 83
BC107328.2 - 3 - 4 649 83

View Full Product Details