Assay Design



Gene Transcripts

Gene Symbol
Osbp
Entrez Gene ID
76303
Gene Name
oxysterol binding protein
Gene Aliases
1110018F06Rik, AW559088, mKIAA4220
Chromosome Location
Chr.19: 11965844 - 11994114 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033174.1 NP_001028346.1 4 - 5 1118 116
GenBank mRNA AK132663.1 - 2 - 3 398 116
AK220301.1 - 1 - 2 156 116
BC107327.1 - 4 - 5 852 116
BC107328.2 - 4 - 5 852 116

View Full Product Details