Assay Design



Gene Transcripts

Gene Symbol
Osbp
Entrez Gene ID
76303
Gene Name
oxysterol binding protein
Gene Aliases
1110018F06Rik, AW559088, mKIAA4220
Chromosome Location
Chr.19: 11965844 - 11994114 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033174.1 NP_001028346.1 5 - 6 1215 68
GenBank mRNA AK132663.1 - 3 - 4 495 68
AK220301.1 - 2 - 3 253 68
BC107327.1 - 5 - 6 949 68
BC107328.2 - 5 - 6 949 68

View Full Product Details