Assay Design



Gene Transcripts

Gene Symbol
Oplah
Entrez Gene ID
75475
Gene Name
5-oxoprolinase (ATP-hydrolysing)
Gene Aliases
1700010G02Rik
Chromosome Location
Chr.15: 76296603 - 76331101 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_153122.2 NP_694762.1 3 - 4 437 85
GenBank mRNA AK158391.1 - 3 - 4 479 85
BC034522.1 - 3 - 4 437 85

View Full Product Details