Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163316.1 | NP_001156788.1 | 7 - 8 | 778 | 61 |
NM_026269.2 | NP_080545.2 | 7 - 8 | 969 | 61 | |
GenBank mRNA | AK003551.1 | - | 7 - 8 | 918 | 61 |
BC019557.1 | - | 7 - 8 | 760 | 61 |