Assay Design



Gene Transcripts

Gene Symbol
Iglon5
Entrez Gene ID
210094
Gene Name
IgLON family member 5
Gene Aliases
A230106M20Rik, Gm1431
Chromosome Location
Chr.7: 43472907 - 43490057 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164518.1 NP_001157990.1 5 - 6 641 107
GenBank mRNA BC116421.1 - 2 - 3 225 107
BC116422.1 - 2 - 3 225 107

View Full Product Details