Assay Design



Gene Transcripts

Gene Symbol
Ptar1
Entrez Gene ID
72351
Gene Name
protein prenyltransferase alpha subunit repeat containing 1
Gene Aliases
1700084D09Rik, 2010013L17Rik, 4930428J16Rik, AI449677
Chromosome Location
Chr.19: 23686898 - 23723092 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_028208.1 NP_082484.1 4 - 5 489 124
GenBank mRNA AK006988.1 - 4 - 5 579 124
AK015225.1 - 4 - 5 557 124

View Full Product Details