Assay Design
Gene Transcripts
- Gene Symbol
- Vmn1r82
- Entrez Gene ID
-
171268
- Gene Name
- vomeronasal 1 receptor 82
- Gene Aliases
-
V1rg12
- Chromosome Location
-
Chr.7: 12304805 - 12305719 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_134234.1 |
NP_598995.1 |
1 - 1 |
875 |
125 |
GenBank mRNA |
BC109325.2 |
- |
1 - 1 |
875 |
125 |
View Full Product Details