Assay Design
Gene Transcripts
- Gene Symbol
- Hrk
- Entrez Gene ID
-
12123
- Gene Name
- harakiri, BCL2 interacting protein (contains only BH3 domain)
- Gene Aliases
-
AI838259, Bid3, DP5, harakiri
- Chromosome Location
-
Chr.5: 118169764 - 118189478 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_007545.2 |
NP_031571.1 |
1 - 2 |
368 |
85 |
GenBank mRNA |
BC128022.1 |
- |
2 - 3 |
189 |
85 |
View Full Product Details