Assay Design



Gene Transcripts

Gene Symbol
Asb5
Entrez Gene ID
76294
Gene Name
ankyrin repeat and SOCs box-containing 5
Gene Aliases
1110018D09Rik
Chromosome Location
Chr.8: 54550331 - 54587842 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029569.3 NP_083845.1 5 - 6 783 67
GenBank mRNA AF398966.1 - 5 - 6 694 67
AK003770.1 - 5 - 6 784 67
BC108405.1 - 5 - 6 698 67
BC120851.1 - 5 - 6 749 67
BC132007.1 - 5 - 6 749 67

View Full Product Details