Assay Design



Gene Transcripts

Gene Symbol
Asb5
Entrez Gene ID
76294
Gene Name
ankyrin repeat and SOCs box-containing 5
Gene Aliases
1110018D09Rik
Chromosome Location
Chr.8: 54550331 - 54587842 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029569.3 NP_083845.1 6 - 7 982 87
GenBank mRNA AF398966.1 - 6 - 7 893 87
AK003770.1 - 6 - 7 983 87
BC108405.1 - 6 - 7 897 87
BC120851.1 - 6 - 7 948 87
BC132007.1 - 6 - 7 948 87

View Full Product Details