Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009580.2 | NP_033606.2 | 11 - 12 | 1761 | 64 |
GenBank mRNA | BC100347.1 | - | 11 - 12 | 1779 | 64 |
BC125613.1 | - | 11 - 12 | 1761 | 64 | |
BC125615.1 | - | 11 - 12 | 1761 | 64 | |
U20448.1 | - | 11 - 12 | 1792 | 64 |