Assay Design
Gene Transcripts
- Gene Symbol
- Dydc1
- Entrez Gene ID
-
69496
- Gene Name
- DPY30 domain containing 1
- Gene Aliases
-
1700029M23Rik
- Chromosome Location
-
Chr.14: 41072911 - 41092197 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_027094.1 |
NP_081370.1 |
3 - 4 |
399 |
106 |
GenBank mRNA |
AK006511.1 |
- |
3 - 4 |
399 |
106 |
View Full Product Details