Assay Design



Gene Transcripts

Gene Symbol
Rfc1
Entrez Gene ID
19687
Gene Name
replication factor C (activator 1) 1
Gene Aliases
140kDa, Alp145, RFC140, Recc1
Chromosome Location
Chr.5: 65261852 - 65336476 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011258.2 NP_035388.2 6 - 7 756 73
GenBank mRNA AK136937.1 - 6 - 7 735 73
AK146164.1 - 6 - 7 751 73
AK146314.1 - 6 - 7 751 73
BC085173.1 - 6 - 7 733 73
BC085174.1 - 6 - 7 725 73
U01222.1 - 6 - 7 724 73
U07157.1 - 6 - 7 722 73
U15037.1 - 6 - 7 757 73
U36441.1 - 6 - 7 733 73
X72711.1 - 6 - 7 722 73

View Full Product Details