Assay Design



Gene Transcripts

Gene Symbol
Arhgef7
Entrez Gene ID
54126
Gene Name
Rho guanine nucleotide exchange factor (GEF7)
Gene Aliases
Cool, PIX, Pak3bp, betaPix, betaPix-b, betaPix-c, cool-1, mKIAA0142, p85Cool1, p85SPR
Chromosome Location
Chr.8: 11728105 - 11835219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113517.1 NP_001106989.1 12 - 13 1566 67
NM_001113518.1 NP_001106990.1 11 - 12 1355 67
NM_017402.4 NP_059098.2 11 - 12 1355 67
GenBank mRNA AF343877.1 - 11 - 12 1235 67
AK052545.1 - 11 - 12 1326 67
AK129064.1 - 12 - 13 1502 67
AK139156.1 - 12 - 13 1495 67
BC044838.1 - 11 - 12 1340 67

View Full Product Details