Assay Design



Gene Transcripts

Gene Symbol
Arhgef7
Entrez Gene ID
54126
Gene Name
Rho guanine nucleotide exchange factor (GEF7)
Gene Aliases
Cool, PIX, Pak3bp, betaPix, betaPix-b, betaPix-c, cool-1, mKIAA0142, p85Cool1, p85SPR
Chromosome Location
Chr.8: 11728105 - 11835219 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001113517.1 NP_001106989.1 13 - 14 1651 68
NM_001113518.1 NP_001106990.1 12 - 13 1440 68
NM_017402.4 NP_059098.2 12 - 13 1440 68
GenBank mRNA AF343877.1 - 12 - 13 1320 68
AK052545.1 - 12 - 13 1411 68
AK129064.1 - 13 - 14 1587 68
AK139156.1 - 13 - 14 1580 68
BC044838.1 - 12 - 13 1425 68

View Full Product Details