Assay Design



Gene Transcripts

Gene Symbol
Setd4
Entrez Gene ID
224440
Gene Name
SET domain containing 4
Gene Aliases
AY037804, C21orf18, ORF21
Chromosome Location
Chr.16: 93583457 - 93604833 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145482.3 NP_663457.2 6 - 7 708 73
GenBank mRNA AY037804.1 - 6 - 7 708 73
BC029101.1 - 6 - 7 605 73
BY327995.1 - 4 - 5 313 73

View Full Product Details