Assay Design



Gene Transcripts

Gene Symbol
Cngb3
Entrez Gene ID
30952
Gene Name
cyclic nucleotide gated channel beta 3
Gene Aliases
CCNC2, CNG6, Cngbeta2
Chromosome Location
Chr.4: 19280850 - 19510623 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013927.2 NP_038955.1 15 - 16 1845 59
GenBank mRNA AJ243572.1 - 15 - 16 1845 59
AK040140.1 - 6 - 7 712 59

View Full Product Details