Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167879.1 | NP_001161351.1 | 2 - 3 | 352 | 104 |
GenBank mRNA | BC060268.1 | - | 1 - 2 | 136 | 104 |
BC113782.1 | - | 1 - 2 | 782 | 104 | |
BC113783.1 | - | 1 - 2 | 783 | 104 |