Assay Design



Gene Transcripts

Gene Symbol
Gm11837
Entrez Gene ID
100038514
Gene Name
predicted gene 11837
Gene Aliases
OTTMUSG00000004551
Chromosome Location
Chr.4: 14929829 - 14953031 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243100.1 NP_001230029.1 3 - 4 308 97
GenBank mRNA AK005641.1 - 2 - 3 95 97


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CB274706.1 - 3 - 4 298 Chr. 4 - 14930651 - 14952970 [+] on Build GRCm38 97
GenBankmRNA BU962870.1 - 3 - 4 269 Chr. 4 - 14930683 - 14952917 [+] on Build GRCm38 97
GenBankmRNA CA464091.1 - 3 - 4 289 Chr. 4 - 14930661 - 14952967 [+] on Build GRCm38 97
GenBankmRNA BU938860.1 - 290 Chr. - - [-] on Build 97
GenBankmRNA CA463133.1 - 3 - 4 281 Chr. 4 - 14930669 - 14952832 [+] on Build GRCm38 97
GenBankmRNA CN839853.1 - 3 - 4 259 Chr. 4 - 14930693 - 14952800 [+] on Build GRCm38 97
GenBankmRNA CD363818.1 - 3 - 4 310 Chr. 4 - 14929908 - 14952893 [+] on Build GRCm38 97
GenBankmRNA AA183467.1 - 3 - 4 244 Chr. 4 - 14930709 - 14947525 [+] on Build GRCm38 97
GenBankmRNA CA463179.1 - 3 - 4 277 Chr. 4 - 14930676 - 14952890 [+] on Build GRCm38 97
GenBankmRNA CN835596.1 - 3 - 4 311 Chr. 4 - 14930641 - 14952760 [+] on Build GRCm38 97
GenBankmRNA CN833750.1 - 3 - 4 279 Chr. 4 - 14930671 - 14952815 [+] on Build GRCm38 97
GenBankmRNA BU938202.1 - 157 Chr. - - [-] on Build 97
GenBankmRNA BU938289.1 - 273 Chr. - - [-] on Build 97


Adjacent Gene Details

Gene Name Gene Symbol Alias Distance Relative Direction Location Chromosome
Upstream transmembrane protein 55A Tmem55a 2610319K07Rik, AI315591, AV001360 -23.2kb Sense Chr. - 14864219 - 14915260 on Build GRCm38
Downstream N-terminal EF-hand calcium binding protein 1 Necab1 1700003H21Rik, Efcbp1, STIP-1 10.0kb Anti-sense Chr. - 14952245 - 15149804 on Build GRCm38

View Full Product Details