Assay Design



Gene Transcripts

Gene Symbol
Nom1
Entrez Gene ID
433864
Gene Name
nucleolar protein with MIF4G domain 1
Gene Aliases
D5Kng1, Gm1040
Chromosome Location
Chr.5: 29434659 - 29452169 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033457.2 NP_001028629.2 7 - 8 2031 110
GenBank mRNA AK148406.1 - 7 - 8 2035 110

View Full Product Details