Assay Design



Gene Transcripts

Gene Symbol
Slc2a9
Entrez Gene ID
117591
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 9
Gene Aliases
Glut9, SLC2A9B, SLC2a9A
Chromosome Location
Chr.5: 38349273 - 38503149 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012363.2 NP_001012363.2 10 - 11 1213 64
NM_001102414.1 NP_001095884.1 10 - 11 1412 64
NM_001102415.1 NP_001095885.1 8 - 9 1091 64
NM_145559.2 NP_663534.1 9 - 10 1086 64
GenBank mRNA AF469480.1 - 10 - 11 1416 64
AF490463.1 - 8 - 9 916 64
AK033725.1 - 3 - 4 283 64
AK037093.1 - 9 - 10 1077 64
AK156740.1 - 10 - 11 1316 64
AK172234.1 - 10 - 11 1316 64
AY776155.1 - 10 - 11 1192 64
BC006076.1 - 9 - 10 1086 64
BC138213.1 - 11 - 12 1348 64
BC145198.1 - 12 - 13 1348 64

View Full Product Details