Assay Design



Gene Transcripts

Gene Symbol
Mcoln1
Entrez Gene ID
94178
Gene Name
mucolipin 1
Gene Aliases
2210015I05Rik, TRPML1, mucolipidin
Chromosome Location
Chr.8: 3500442 - 3515232 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_053177.1 NP_444407.1 3 - 4 481 66
GenBank mRNA AF302009.1 - 3 - 4 478 66
AK028385.1 - 3 - 4 458 66
AK034471.1 - 3 - 4 492 66
AK038594.1 - 3 - 4 494 66
AK049606.1 - 3 - 4 482 66
BC005651.1 - 3 - 4 481 66
BC020377.1 - 2 - 3 331 66

View Full Product Details