Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016897.3 | NP_058593.2 | 6 - 7 | 658 | 61 |
GenBank mRNA | AB021122.1 | - | 6 - 7 | 638 | 61 |
AK013976.1 | - | 6 - 7 | 604 | 61 | |
AK146516.1 | - | 6 - 7 | 624 | 61 | |
BC037022.1 | - | 7 - 8 | 635 | 61 | |
BC062963.1 | - | 6 - 7 | 594 | 61 | |
BC069903.1 | - | 6 - 7 | 628 | 61 | |
BC096669.1 | - | 6 - 7 | 627 | 61 | |
BC132019.1 | - | 6 - 7 | 607 | 61 | |
BC132319.1 | - | 6 - 7 | 607 | 61 |