Assay Design
Gene Transcripts
- Gene Symbol
- Insr
- Entrez Gene ID
-
16337
- Gene Name
- insulin receptor
- Gene Aliases
-
4932439J01Rik, CD220, D630014A15Rik, IR, IR-A, IR-B
- Chromosome Location
-
Chr.8: 3150922 - 3279649 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_010568.2 |
NP_034698.2 |
18 - 19 |
3985 |
79 |
GenBank mRNA |
J05149.1 |
- |
18 - 19 |
3544 |
79 |
KC356871.1 |
- |
19 - 20 |
3532 |
79 |
View Full Product Details