Assay Design



Gene Transcripts

Gene Symbol
Nfxl1
Entrez Gene ID
100978
Gene Name
nuclear transcription factor, X-box binding-like 1
Gene Aliases
1700012H24Rik, AW538212, D430033A06Rik, DNABF, GCF, Gm1058, TCF9
Chromosome Location
Chr.5: 72513301 - 72559707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133921.2 NP_598682.2 22 - 23 2637 68
GenBank mRNA AK132320.1 - 21 - 22 2547 68
BC022652.1 - 6 - 7 637 68
BC032981.1 - 2 - 3 320 68

View Full Product Details