Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011669.3 | NP_035799.1 | 4 - 5 | 868 | 106 |
GenBank mRNA | AF022792.1 | - | 1 - 2 | 137 | 106 |
AF441835.1 | - | 3 - 4 | 529 | 106 | |
AK006739.1 | - | 4 - 5 | 918 | 106 | |
AK035629.1 | - | 4 - 5 | 709 | 106 | |
AK167153.1 | - | 4 - 5 | 708 | 106 | |
BC049274.1 | - | 2 - 3 | 535 | 106 | |
BC068136.1 | - | 4 - 5 | 868 | 106 |