Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011669.3 | NP_035799.1 | 8 - 9 | 1307 | 105 |
GenBank mRNA | AF441835.1 | - | 7 - 8 | 968 | 105 |
AK006739.1 | - | 8 - 9 | 1357 | 105 | |
AK035629.1 | - | 8 - 9 | 1148 | 105 | |
AK167153.1 | - | 8 - 9 | 1147 | 105 | |
BC023858.1 | - | 2 - 3 | 140 | 105 | |
BC049274.1 | - | 6 - 7 | 974 | 105 | |
BC055398.1 | - | 4 - 5 | 399 | 105 | |
BC068136.1 | - | 8 - 9 | 1307 | 105 |