Assay Design
Gene Transcripts
- Gene Symbol
- Lgi2
- Entrez Gene ID
-
246316
- Gene Name
- leucine-rich repeat LGI family, member 2
- Gene Aliases
-
mKIAA1916
- Chromosome Location
-
Chr.5: 52533517 - 52566462 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK122570.1 |
- |
4 - 5 |
539 |
143 |
AY841361.1 |
- |
4 - 5 |
399 |
143 |
View Full Product Details