Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025951.3 | NP_080227.2 | 2 - 3 | 539 | 89 |
GenBank mRNA | AF411321.1 | - | 2 - 3 | 394 | 89 |
AK011751.1 | - | 2 - 3 | 502 | 89 | |
AK035523.1 | - | 2 - 3 | 524 | 89 | |
AK170730.1 | - | 2 - 3 | 523 | 89 | |
BC062144.1 | - | 2 - 3 | 446 | 89 |