Assay Design



Gene Transcripts

Gene Symbol
Dok7
Entrez Gene ID
231134
Gene Name
docking protein 7
Gene Aliases
A930013K19Rik, AW049091, Dok-7, EF-12, Oit5
Chromosome Location
Chr.5: 35056566 - 35087839 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172708.3 NP_766296.1 2 - 3 284 70
GenBank mRNA AB220919.1 - 3 - 4 330 70
AK044445.1 - 2 - 3 285 70
BC089590.1 - 3 - 4 403 70

View Full Product Details