Assay Design



Gene Transcripts

Gene Symbol
Sult1e1
Entrez Gene ID
20860
Gene Name
sulfotransferase family 1E, member 1
Gene Aliases
EST, ST1E1, Ste
Chromosome Location
Chr.5: 87575968 - 87591611 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023135.2 NP_075624.2 5 - 6 635 145
GenBank mRNA AK015750.1 - 5 - 6 619 145
BC034891.1 - 5 - 6 576 145
S78182.1 - 5 - 6 618 145

View Full Product Details