Assay Design



Gene Transcripts

Gene Symbol
Plrg1
Entrez Gene ID
53317
Gene Name
pleiotropic regulator 1, PRL1 homolog (Arabidopsis)
Gene Aliases
AA958940, C80566, Tango4
Chromosome Location
Chr.3: 83055538 - 83072291 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016784.3 NP_058064.2 14 - 15 1593 103
GenBank mRNA AF044334.1 - 13 - 14 1485 103
AK075993.1 - 14 - 15 1581 103
AK076007.1 - 14 - 15 1571 103
AK077197.1 - 14 - 15 1594 103
AK080068.1 - 14 - 15 1574 103
BC006750.1 - 14 - 15 1563 103
BC052895.1 - 14 - 15 1551 103

View Full Product Details