Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016784.3 | NP_058064.2 | 9 - 10 | 939 | 69 |
GenBank mRNA | AF044334.1 | - | 8 - 9 | 831 | 69 |
AK075993.1 | - | 9 - 10 | 927 | 69 | |
AK076007.1 | - | 9 - 10 | 917 | 69 | |
AK077197.1 | - | 9 - 10 | 940 | 69 | |
AK080068.1 | - | 9 - 10 | 946 | 69 | |
BC006750.1 | - | 9 - 10 | 909 | 69 | |
BC052895.1 | - | 9 - 10 | 897 | 69 |