Assay Design



Gene Transcripts

Gene Symbol
Prkcd
Entrez Gene ID
18753
Gene Name
protein kinase C, delta
Gene Aliases
AI385711, D14Ertd420e, PKC[d], PKCdelta, Pkcd
Chromosome Location
Chr.14: 30595353 - 30626414 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011103.3 NP_035233.1 11 - 12 1322 90
GenBank mRNA AB011812.1 - 10 - 11 1163 90
AB201453.1 - 8 - 9 924 90
AB201454.1 - 8 - 9 1002 90
AB201455.1 - 8 - 9 924 90
AB201456.1 - 8 - 9 1002 90
AF251036.1 - 10 - 11 1085 90
AK144231.1 - 11 - 12 1327 90
AK168768.1 - 11 - 12 1317 90
AY545076.1 - 10 - 11 1085 90
BC051416.1 - 11 - 12 1286 90
M69042.1 - 11 - 12 1317 90
X60304.1 - 10 - 11 1098 90

View Full Product Details