Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165930.1 | NP_001159402.1 | 6 - 7 | 613 | 89 |
NM_029626.2 | NP_083902.2 | 6 - 7 | 1142 | 89 | |
GenBank mRNA | AK077388.1 | - | 6 - 7 | 606 | 89 |
AK160461.1 | - | 7 - 8 | 773 | 89 | |
BC029215.1 | - | 4 - 5 | 816 | 89 | |
BC069873.1 | - | 6 - 7 | 692 | 89 | |
CX209391.1 | - | 5 - 6 | 573 | 89 |