Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001164359.1 | NP_001157831.1 | 4 - 5 | 439 | 114 |
NM_001164360.1 | NP_001157832.1 | 4 - 5 | 347 | 114 | |
NM_145502.3 | NP_663477.3 | 3 - 4 | 543 | 114 | |
GenBank mRNA | AK133557.1 | - | 3 - 4 | 286 | 114 |
AK145990.1 | - | 4 - 5 | 320 | 114 | |
BC011220.1 | - | 4 - 5 | 311 | 114 | |
BC023849.1 | - | 3 - 4 | 406 | 114 | |
CJ134812.1 | - | 4 - 5 | 420 | 114 |