Assay Design
Gene Transcripts
- Gene Symbol
- Ythdf1
- Entrez Gene ID
-
228994
- Gene Name
- YTH domain family 1
- Gene Aliases
-
2210410K23Rik, 8030473O16
- Chromosome Location
-
Chr.2: 180904377 - 180920936 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK033241.1 |
- |
3 - 4 |
374 |
73 |
View Full Product Details